6-Acetyl-2-Benzoxazolinone ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 6-Acetyl-2-benzoxazolinone, 6-Acetyl-2(3H)-benzoxazolone, 6-Acetyl-2-hydroxybenzoxazole, 6-Acetyl-2-benzoxazolone, 6-Acetylbenzo[d]oxazol-2(3H)-one

A151748-1G A151748-250MG A151748-25G A151748-5G
ALA151748-1GEA 64.51 USD
ALA151748-1G ALA151748250MG ALA151748-25G ALA151748-5G
6-Acetyl-2-Benzoxazolinone ≥98% (by HPLC)
6-Acetyl-2-Benzoxazolinone
Formula: C₉H₇NO₃
Melting Pt: 236 °C
MDL Number: MFCD01664312
CAS Number: 54903-09-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=215b71146af10b0fe50925fe883e79f8c3707b76&vid=215b71146af10b0fe50925fe883e79f8c3707b76&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR