2-Amino-3-bromo-5,6-dimethylpyridine ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A151747-1G A151747-250MG A151747-5G
ALA151747-1GEA 117.71 USD
ALA151747-1G ALA151747250MG ALA151747-5G
2-Amino-3-bromo-5,6-dimethylpyridine ≥98%
2-Amino-3-bromo-5,6-dimethylpyridine
Formula: C₇H₉BrN₂
Melting Pt: 114.0 to 118.0 °C
MDL Number: MFCD04039386
CAS Number: 161091-49-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ddd5039cb020cdd5f2a883a4c9b91180c58a0627&vid=ddd5039cb020cdd5f2a883a4c9b91180c58a0627&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR