2-Amino-5-iodo-4-methylpyridine ≥98% (by GC, titration analysis)
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Danger
|
Synonyms:
5-Iodo-4-methyl-pyridin-2-ylamine, 5-Iodo-4-methylpyridin-2-ylamine
A151744-10G
A151744-1G
A151744-250MG
ALA151744-10GEA
360.91
USD
ALA151744-10G
ALA151744-1G
ALA151744250MG
2-Amino-5-iodo-4-methylpyridine ≥98% (by GC, titration analysis)
2-Amino-5-iodo-4-methylpyridine
Formula:
C₆H₇IN₂ MW: 234.04 g/mol Melting Pt: 119.0 - 123.0 °C |
MDL Number:
MFCD06637727 CAS Number: 356561-08-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...