6-Acrylamidohexanoic acid 98 titration

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 6-Acrylamidohexanoic Acid

A151732-1G A151732-200MG A151732-5G
ALA151732-1GEA 80.66 USD
ALA151732-1G ALA151732200MG ALA151732-5G
6-Acrylamidohexanoic acid 98 titration
6-Acrylamidohexanoic acid
Formula: C₉H₁₅NO₃
MW: 185.22 g/mol
Melting Pt: 89 °C
MDL Number: MFCD06804452
CAS Number: 20766-85-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e7b86e1896e596142c48758dd94b767f8061b43&vid=5e7b86e1896e596142c48758dd94b767f8061b43&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR