1-(2-Aminoethyl)-4-methylpiperazine hydrochloride ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 2-(4-Methyl-1-piperazino)ethylamine Hydrochloride, 1-(2-Aminoethyl)-4-methylpiperazine Hydrochloride

A151725-1G A151725-200MG A151725-5G
ALA151725-1GEA 210.81 USD
ALA151725-1G ALA151725200MG ALA151725-5G
1-(2-Aminoethyl)-4-methylpiperazine hydrochloride ≥98%
1-(2-Aminoethyl)-4-methylpiperazine hydrochloride
Formula: C₇H₁₇N₃
MW: 143 g/mol (free)
CAS Number: 646051-17-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e3baf3a3ebeaeabcd2bfc4c580e1cb7da56528c7&vid=e3baf3a3ebeaeabcd2bfc4c580e1cb7da56528c7&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR