1-Amino-4-methylpiperazine ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 4-Methylpiperazin-1-amine

A151724-100ML A151724-25ML A151724-5ML
ALA151724100MLEA 252.61 USD
ALA151724100ML ALA151724-25ML ALA151724-5ML
1-Amino-4-methylpiperazine ≥98% (by GC)
1-Amino-4-methylpiperazine

1-Amino-4-methylpiperazine has been used:in synthesis of pyrazolo-pyrazine, -pyridine derivatives and 2-[(4-methylpiperazin-1-yl)iminomethyl]phenol;as a solvating and stabilizing agent in the preparation of uniform Ag nanoparticles (<10nm).

Formula: C₅H₁₃N₃
MW: 115.18 g/mol
Boiling Pt: 175°C
Density: 0.963 g/cm³
Flash Pt: 103°C
Storage Temperature: Ambient
MDL Number: MFCD00006154
CAS Number: 6928-85-4
EINECS: 230-053-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=013ce0db3657b80011c40403c8799dc53e6d9956&vid=013ce0db3657b80011c40403c8799dc53e6d9956&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR