(±)-2-Aminooctane ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 2-Aminooctane, (±)-2-Octylamine, (±)-1-Methylheptylamine, 1-Methylheptylamine, sec-Octylamine

A151716-1G A151716-25G A151716-5G
ALA151716-1GEA 60.71 USD
ALA151716-1G ALA151716-25G ALA151716-5G
(±)-2-Aminooctane ≥98%
(±)-2-Aminooctane
Formula: C₈H₁₉N
MW: 129.25 g/mol
Boiling Pt: 165°C
Density: 0.77 g/cm³
Flash Pt: 50 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008103
CAS Number: 693-16-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1941ec874cd33a3a98d3343675df0e152249ea55&vid=1941ec874cd33a3a98d3343675df0e152249ea55&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR