3-Acetyl-6-bromo-2H-chromen-2-one ≥96%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 3-Acetyl-6-bromocoumarin, 3-Acetyl-6-bromo-2H-chromen-2-one

A151701-1G A151701-250MG A151701-5G
ALA151701-1GEA 79.71 USD
ALA151701-1G ALA151701250MG ALA151701-5G
3-Acetyl-6-bromo-2H-chromen-2-one ≥96%
3-Acetyl-6-bromo-2H-chromen-2-one
Formula: C₁₁H₇BrO₃
MW: 267.09 g/mol
Melting Pt: 225 °C
MDL Number: MFCD00024075
CAS Number: 2199-93-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8028bf57e9dbd385b1b74a2bc8edb4b70ffed0f&vid=b8028bf57e9dbd385b1b74a2bc8edb4b70ffed0f&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR