4-[2-(2-Amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoic acid hydrate ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 4-[2-(2-Amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoic Acid, 4-(2-(2-Amino-4-oxo-4,7-dihydro-1H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl)benzoic acid, 2-Amino-4,7-dihydro-5-[2-[4-(carboxy)phenyl]ethyl]-4-oxo-3H-pyrrolo[2,3-d]pyrimidine

A151697-1G A151697-250MG A151697-25G A151697-5G
ALA151697-1GEA 35.06 USD
ALA151697-1G ALA151697250MG ALA151697-25G ALA151697-5G
4-[2-(2-Amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoic acid hydrate ≥98%
4-[2-(2-Amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoic acid hydrate
Formula: C₁₅H₁₄N₄O₃
MW: 298.30 g/mol
MDL Number: MFCD11040439
CAS Number: 137281-39-1
EINECS: 429-790-9, 682-744-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9542169739d8114718f8e20a73d3948e138baf17&vid=9542169739d8114718f8e20a73d3948e138baf17&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR