4-Amino-2-methoxybenzonitrile ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A151696-1G A151696-250MG
ALA151696-1GEA 122.46 USD
ALA151696-1G ALA151696250MG
4-Amino-2-methoxybenzonitrile ≥97%
4-Amino-2-methoxybenzonitrile
Formula: C₈H₈N₂O
Melting Pt: 100 °C
MDL Number: MFCD09027303
CAS Number: 7251-09-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR