2-(4-Amino-2-nitroanilino)ethanol ≥95% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 4-Amino-N-(2-hydroxyethyl)-2-nitroaniline, N1-(2-Hydroxyethyl)-2-nitro-1,4-phenylenediamine, 2-((4-Amino-2-nitrophenyl)amino)ethanol

A151695-100G A151695-25G A151695-5G
ALA151695-100GEA 718.11 USD
ALA151695-100G ALA151695-25G ALA151695-5G
2-(4-Amino-2-nitroanilino)ethanol ≥95% (by HPLC)
2-(4-Amino-2-nitroanilino)ethanol
Formula: C₈H₁₁N₃O₃
Melting Pt: 126 °C
MDL Number: MFCD00071801
CAS Number: 2871-01-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR