2'-Amino-3'-hydroxyacetophenone ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 1-(2-Amino-3-hydroxyphenyl)ethanone

A151688-1G A151688-250MG A151688-5G
ALA151688-1GEA 44.56 USD
ALA151688-1G ALA151688250MG ALA151688-5G
2'-Amino-3'-hydroxyacetophenone ≥98%
2'-Amino-3'-hydroxyacetophenone
Formula: C₈H₉NO₂
MW: 151.1626 g/mol
Melting Pt: 182°C
Storage Temperature: Ambient
MDL Number: MFCD00191301
CAS Number: 4502-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR