4-Amino-2,3-xylenol ≥98% (by GC, titration analysis)

Supplier: ALADDIN SCIENTIFIC
Warning

A151686-10G A151686-1G
ALA151686-10GEA 349.51 USD
ALA151686-10G ALA151686-1G
4-Amino-2,3-xylenol ≥98% (by GC, titration analysis)
4-Amino-2,3-xylenol
Formula: C₈H₁₁NO
Melting Pt: 174 °C
Storage Temperature: Ambient
MDL Number: MFCD00070554
CAS Number: 3096-69-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eff632cbf934a3eea7acbb09aa3303caee8f2e2f&vid=eff632cbf934a3eea7acbb09aa3303caee8f2e2f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR