4'-Aminoacetophenone hydrochloride ≥98%
Supplier: ALADDIN SCIENTIFIC
|
Danger
|
Synonyms:
4'-Aminoacetophenone Hydrochloride, 1-(4-Aminophenyl)ethanone hydrochloride
A151685-25G
A151685-5G
ALA151685-25GEA
241.21
USD
ALA151685-25G
ALA151685-5G
4'-Aminoacetophenone hydrochloride ≥98%
4'-Aminoacetophenone hydrochloride
Formula:
C₈H₁₀ClNO |
MDL Number:
MFCD00060232 CAS Number: 41784-08-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...