α-Cyclopropylbenzyl alcohol ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: alpha-Cyclopropylbenzyl alcohol, Cyclopropyl(phenyl)methanol, α-Cyclopropylbenzyl alcohol, Cyclopropyl phenyl carbinol, Cyclopropylphenylcarbinol

A151682-5ML A151682-1ML A151682-25ML
ALA151682-5MLEA 101.56 USD
ALA151682-5ML ALA151682-1ML ALA151682-25ML
α-Cyclopropylbenzyl alcohol ≥95%
α-Cyclopropylbenzyl alcohol
Formula: C₁₀H₁₂O
Boiling Pt: 132°C
Density: 1.037 g/cm³
Flash Pt: 112°C
MDL Number: MFCD00001299
CAS Number: 1007-03-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c49fb08ab20fb04ff441f02df69bd3239d8bbd5e&vid=c49fb08ab20fb04ff441f02df69bd3239d8bbd5e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR