2-Amino-5-bromo-3-methylpyridine ≥98%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
A151675-5G
A151675-100G
A151675-1G
A151675-25G
ALA151675-5GEA
81.61
USD
ALA151675-5G
ALA151675-100G
ALA151675-1G
ALA151675-25G
2-Amino-5-bromo-3-methylpyridine ≥98%
2-Amino-5-bromo-3-methylpyridine
2-Amino-5-bromo-3-methylpyridine can be prepared from 2-amino-3-methylpyridine, via bromination.2-Amino-5-bromo-3-methylpyridine may be used to synthesize:5-bromo-2-chloro-3-methylpyridine;2-azidopyridine 1-oxide;2-amino-5-bromo-3-methylpyridine 1-oxide;2,5-dibromo-3-methyl pyridine;ethyl-3,6-dibromo-8-methylimidazo[1,2-a]pyridine-2-carboxylate.
Formula:
C₆H₇BrN₂ Melting Pt: 94 °C |
MDL Number:
MFCD00068232 CAS Number: 3430-21-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...