2-Amino-5-chloro-3-methylpyridine ≥98% (by GC)

Supplier: Aladdin Scientific
Warning

A151671-1G A151671-5G
ALA151671-1GEA 53.11 USD
ALA151671-1G ALA151671-5G
2-Amino-5-chloro-3-methylpyridine ≥98% (by GC)
2-Amino-5-chloro-3-methylpyridine
Formula: C₆H₇ClN₂
Melting Pt: 70 °C
MDL Number: MFCD00129030
CAS Number: 20712-16-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9761d64cfc0aef38af70e65bda20ab43f3575e08&vid=9761d64cfc0aef38af70e65bda20ab43f3575e08&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR