2-Amino-3-bromo-5-methylpyridine ≥98%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
2-Amino-3-bromo-5-methylpyridine may be used in the synthesis of: . 3-bromo-2-chloro-5-methylpyridine . 3-bromo-2-fluoro-5-methylpyridine . 8-methyl-1,2,3,5-tetrahydropyrido[3,4-b][1,4]diazepin-4-one . 3-bromo-2-(ethylamino)-5-methylpyridine . (3-bromo-5-methylpyridin-2-yl)aniline . 5-methyl-3-(phenylethynyl)pyridin-2-amine . 3-[(2-methoxyphenyl)ethynyl]-5-methylpyridin-2-amine . 3-[(4-methoxyphenyl)ethynyl]-5-methylpyridin-2-amine . 3-[(3-chlorophenyl)ethynyl]-5-methylpyridin-2-amine
Formula:
C₆H₇BrN₂ Melting Pt: 73-76°C |
MDL Number:
MFCD00068231 CAS Number: 17282-00-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...