4'-Amino-3',5'-dichloroacetophenone ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

A151669-100G A151669-25G A151669-5G
ALA151669-100GEA 566.11 USD
ALA151669-100G ALA151669-25G ALA151669-5G
4'-Amino-3',5'-dichloroacetophenone ≥98% (by GC)
4'-Amino-3',5'-dichloroacetophenone
Formula: C₈H₇Cl₂NO
Melting Pt: 165 °C
Storage Temperature: Ambient
MDL Number: MFCD00238535
CAS Number: 37148-48-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3943ea3d21695dc09d9e3222a5e4251bf7f9bd7f&vid=3943ea3d21695dc09d9e3222a5e4251bf7f9bd7f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR