2-Amino-4-methoxypyridine ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A151667-25G A151667-5G A151667-100G A151667-1G
ALA151667-25GEA 913.81 USD
ALA151667-25G ALA151667-5G ALA151667-100G ALA151667-1G
2-Amino-4-methoxypyridine ≥98%
2-Amino-4-methoxypyridine
Formula: C₆H₈N₂O
MW: 124.14 g/mol
Melting Pt: 118 °C
Storage Temperature: Ambient
MDL Number: MFCD07437849
CAS Number: 10201-73-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2d18082b6154ca4a7ed249cf479a37d9e71d4547&vid=2d18082b6154ca4a7ed249cf479a37d9e71d4547&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR