4-Amino-3-nitropyridine ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 3-Nitropyridin-4-amine, 3-Nitro-4-pyridylamine, 3-Nitropyridin-4-ylamine

A151665-100G A151665-1G A151665-25G A151665-5G
ALA151665-100GEA 838.76 USD
ALA151665-100G ALA151665-1G ALA151665-25G ALA151665-5G
4-Amino-3-nitropyridine ≥98% (by GC)
4-Amino-3-nitropyridine
Formula: C₅H₅N₃O₂
MW: 139.11 g/mol
Melting Pt: 203-207°C
MDL Number: MFCD00160304
CAS Number: 1681-37-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d82826f7f49158e371d4058391e84d35b6ac8b56&vid=d82826f7f49158e371d4058391e84d35b6ac8b56&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR