2-Amino-5-nitro-3-picoline ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 2-Amino-3-methyl-5-nitropyridine, 3-Methyl-5-nitro-2-pyridylamine

A151663-1G A151663-5G
ALA151663-1GEA 24.61 USD
ALA151663-1G ALA151663-5G
2-Amino-5-nitro-3-picoline ≥98%
2-Amino-5-nitro-3-picoline
Formula: C₆H₇N₃O₂
MW: 153.14 g/mol
Boiling Pt: 261 °C
Melting Pt: 255°C
MDL Number: MFCD03095060
CAS Number: 18344-51-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c132d6b1cd4a605aa01b38e76e64468712fd465f&vid=c132d6b1cd4a605aa01b38e76e64468712fd465f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR