Acetoxy-2-propanone ≥97% (by GC)

Supplier: ALADDIN SCIENTIFIC CORPORATION

A151657-25ML A151657-5ML
ALA151657-25MLEA 107.9 USD
ALA151657-25ML ALA151657-5ML
Acetoxy-2-propanone ≥97% (by GC)
Acetoxy-2-propanone
Formula: C₅H₈O₃
Boiling Pt: 174°C
Density: 1.077 g/cm³
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00042848
CAS Number: 592-20-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce1e15c43701626edbb36ce039a46e70e253e48c&vid=ce1e15c43701626edbb36ce039a46e70e253e48c&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR