2,1-Benzisoxazole ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: Anthranil, Benzo[c]isoxazole, 2,1-Benzisoxazole

A151655-1G A151655-5G
ALA151655-1GEA 68.31 USD
ALA151655-1G ALA151655-5G
2,1-Benzisoxazole ≥98% (by GC)
2,1-Benzisoxazole
Formula: C₇H₅NO
MW: 119.1207 g/mol
Boiling Pt: 101°C
Density: 1.19 g/cm³
Flash Pt: 94°C
Storage Temperature: Ambient
MDL Number: MFCD00005806
CAS Number: 271-58-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e7a3f3ccfcfac787b2fee6cbe9c1e4b47979307b&vid=e7a3f3ccfcfac787b2fee6cbe9c1e4b47979307b&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR