3-Amino-5-methylisoxazole ≥97% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 5-Methylisoxazol-3-amine, 3-Amino-5-methylisoxazol, 5-Methylisoxazol-3-ylamine

A151654-100G A151654-25G A151654-5G
ALA151654-100GEA 159.51 USD
ALA151654-100G ALA151654-25G ALA151654-5G
3-Amino-5-methylisoxazole ≥97% (by GC)
3-Amino-5-methylisoxazole

Product Introduction:3-amino-5-methylisoxazole is a major intermediate formed during the biodegradation of Sulfamethoxazole by P. psychrophilicus strain HA-4.It is an intermediate formed during the photocatalytic degradation of sulfamethoxazole (SMX).Application3-amino-5-methylisoxazole for synthesis:• naphtho [1,2-e] [1,3] oxazines• 1-aryl-4-methyl-3,6-bis - (5-methylisoxazol-3-yl) - 2-thio-2,3,6,10 B - tetrahydro-1 H - pyrimido [5,4 - C] quinolin-5-one series with potential mosquito killing activity• hydroxylamine with sulfadiazine and thiamethoxazole

Formula: C₄H₆N₂O
MW: 98.1 g/mol
Boiling Pt: 194
Melting Pt: 60°C
MDL Number: MFCD00003155
CAS Number: 1072-67-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a532785efad86f648ae30804c9a88820c032db2b&vid=a532785efad86f648ae30804c9a88820c032db2b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR