2-Acetoxybenzonitrile ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A151646-100G A151646-1G A151646-25G A151646-5G
ALA151646-100GEA 370.41 USD
ALA151646-100G ALA151646-1G ALA151646-25G ALA151646-5G
2-Acetoxybenzonitrile ≥97%
2-Acetoxybenzonitrile
Formula: C₉H₇NO₂
Boiling Pt: 254°C
Density: 1.14 g/cm³
Flash Pt: 132 °C
MDL Number: MFCD00017360
CAS Number: 5715-02-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR