2'-Amino-4',5'-dimethoxyacetophenone ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A151645-1G A151645-250MG A151645-200MG
ALA151645-1GEA 30.31 USD
ALA151645-1G ALA151645250MG ALA151645-200MG
2'-Amino-4',5'-dimethoxyacetophenone ≥98%
2'-Amino-4',5'-dimethoxyacetophenone
Formula: C₁₀H₁₃NO₃
Melting Pt: 104-108°C
Storage Temperature: Ambient
MDL Number: MFCD00016646
CAS Number: 4101-30-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0cc919b135e6bb8eea67d9e12ce19a0696d499b9&vid=0cc919b135e6bb8eea67d9e12ce19a0696d499b9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR