6-Amino-1,3-benzoxazol-2(3H)-one ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 6-Amino-2-hydroxybenzoxazole, 6-Amino-2-benzoxazolone, 6-Amino-2-benzoxazolinone

A151642-1G A151642-250MG A151642-5G
ALA151642-1GEA 67.36 USD
ALA151642-1G ALA151642250MG ALA151642-5G
6-Amino-1,3-benzoxazol-2(3H)-one ≥97%
6-Amino-1,3-benzoxazol-2(3H)-one
Formula: C₇H₆N₂O₂
MW: 150.14 g/mol
Melting Pt: 205°C
Storage Temperature: Refrigerator
MDL Number: MFCD00463899
CAS Number: 22876-17-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=29930a552a6d3f5bf5f461fe3f5b37ea1eacf6e7&vid=29930a552a6d3f5bf5f461fe3f5b37ea1eacf6e7&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR