2-Bromo-4-(trifluoromethyl)aniline ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 4-Amino-3-bromobenzotrifluoride, 4-Amino-3-bromo-α,α,α-trifluorotoluene, 2-Bromo-α,α,α-trifluoro-p-toluidine

A151638-100G A151638-25G A151638-5G
ALA151638-100GEA 1263.41 USD
ALA151638-100G ALA151638-25G ALA151638-5G
2-Bromo-4-(trifluoromethyl)aniline ≥98% (by GC)
2-Bromo-4-(trifluoromethyl)aniline
Formula: C₇H₅BrF₃N
MW: 240.02 g/mol
Boiling Pt: 109-110 °C
Melting Pt: 32.0-36.0 °C
Density: 1.7 g/cm³
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00042150
CAS Number: 57946-63-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c55b5da1378d172b6572675b1b56714c7ef93288&vid=c55b5da1378d172b6572675b1b56714c7ef93288&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR