(±)-α-Terpineol ≥80% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: α-Terpineol, Terpineol, 2-(4-Methylcyclohex-3-en-1-yl)propan-2-ol

A151627-100ML A151627-25ML
ALA151627100MLEA 61.66 USD
ALA151627100ML ALA151627-25ML
(±)-α-Terpineol ≥80% (by GC)
(±)-α-Terpineol

α-Terpineol is a monoterpene alcohol. It is one of the components responsible for the antifungal activity in Melaleuca alternifolia (tea tree) essential oil.[1] The reaction rate constant of α-terpineol with the OH radical and ozone was found to be (1.9±0.5)×10-10cm3 molecule-1s-1 and (3.0±0.2)×10-16cm3 molecule-1s-1, respectively.

Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Boiling Pt: 210-219℃
Melting Pt: 31-32°C
Flash Pt: 90°C
MDL Number: MFCD00001557
CAS Number: 98-55-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a4c23a9af26fd1d05a618ac1990493922d64db2e&vid=a4c23a9af26fd1d05a618ac1990493922d64db2e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR