5-Amino-2,4,6-triiodoisophthaloyl Dichloride ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A151624-1G A151624-5G
ALA151624-1GEA 24.61 USD
ALA151624-1G ALA151624-5G
5-Amino-2,4,6-triiodoisophthaloyl Dichloride ≥98%
5-Amino-2,4,6-triiodoisophthaloyl Dichloride
Formula: C₈H₂Cl₂I₃NO₂
Melting Pt: 231 °C
Storage Temperature: Ambient
MDL Number: MFCD00270836
CAS Number: 37441-29-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9158f71a172ec7ac9735dd79fe728648608f484&vid=a9158f71a172ec7ac9735dd79fe728648608f484&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR