4-(Aminomethyl)tetrahydropyran ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 4-(Aminomethyl)tetrahydro-2H-pyran, 4-Aminomethyltetrahydropyran, Tetrahydro-2H-pyran-4-ylmethanamine, (Tetrahydro-2H-pyran-4-yl)methanamine

A151618-100G A151618-1G A151618-250MG A151618-5G
ALA151618-100GEA 3105.46 USD
ALA151618-100G ALA151618-1G ALA151618250MG ALA151618-5G
4-(Aminomethyl)tetrahydropyran ≥98% (by GC)
4-(Aminomethyl)tetrahydropyran
Formula: C₆H₁₃NO
Boiling Pt: 76°C
Density: 0.97 g/cm³
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD02179435
CAS Number: 130290-79-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2be0523c5776a250e7109c76071a9c1fc46f299&vid=e2be0523c5776a250e7109c76071a9c1fc46f299&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR