3-Aminobenzenesulfonamide ≥98% (by HPLC, titration analysis)
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
A151617-100G
A151617-25G
A151617-500G
A151617-5G
ALA151617-100GEA
207.96
USD
ALA151617-100G
ALA151617-25G
ALA151617-500G
ALA151617-5G
3-Aminobenzenesulfonamide ≥98% (by HPLC, titration analysis)
3-Aminobenzenesulphonamide
Formula:
C₆H₈N₂O₂S MW: 172.21 g/mol Melting Pt: 139-144 °C |
MDL Number:
MFCD00035781 CAS Number: 98-18-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...