4-Amino-6-chlorobenzene-1,3-disulfonamide ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

A151613-100G A151613-25G A151613-5G
ALA151613-100GEA 120.56 USD
ALA151613-100G ALA151613-25G ALA151613-5G
4-Amino-6-chlorobenzene-1,3-disulfonamide ≥98%
4-Amino-6-chlorobenzene-1,3-disulphonamide

Product Describtion:4-Amino-6-chloro-1,3-benzenedisulfonamide is a potent inhibitor of gastric pathogen, Helicobacter pylori (hpCA).Product Application:4-Amino-6-chloro-1,3-benzenedisulfonamide was used in the synthesis of deuterated thiazides by undergoing condensation with appropriately labeled aldehydes.

Formula: C₆H₈ClN₃O₄S₂
Melting Pt: 250-252°C
Storage Temperature: Ambient
MDL Number: MFCD00007933
CAS Number: 121-30-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a57f76a8730973e910d30e8cc89eab3fba16a119&vid=a57f76a8730973e910d30e8cc89eab3fba16a119&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR