3-Amino-1-benzylpiperidine ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A151611-1G A151611-5G
ALA151611-1GEA 72.11 USD
ALA151611-1G ALA151611-5G
3-Amino-1-benzylpiperidine ≥98%
3-Amino-1-benzylpiperidine
Formula: C₁₂H₁₈N₂
Boiling Pt: 82°C
Storage Temperature: Refrigerator
MDL Number: MFCD04038444
CAS Number: 60407-35-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84dea5ab6ed716a02c0a1171c3404e7c08ed3132&vid=84dea5ab6ed716a02c0a1171c3404e7c08ed3132&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR