4-(2-Aminoethyl)-1-benzylpiperidine ≥98% (by GC, titration analysis)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 2-(1-Benzyl-4-piperidyl)ethylamine

A151606-1G A151606-5G
ALA151606-1GEA 50.26 USD
ALA151606-1G ALA151606-5G
4-(2-Aminoethyl)-1-benzylpiperidine ≥98% (by GC, titration analysis)
4-(2-Aminoethyl)-1-benzylpiperidine
Formula: C₁₄H₂₂N₂
MW: 218.34 g/mol
Density: 0.99 g/cm³
Flash Pt: 136 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02093423
CAS Number: 86945-25-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8c983c4e13122bbfbfcf6ac41f4b5d07a52b3c4&vid=f8c983c4e13122bbfbfcf6ac41f4b5d07a52b3c4&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR