4-(4-ɑ-Cumylphenoxy)phthalonitrile ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 4-(4-(2-Phenylpropan-2-yl)phenoxy)phthalonitrile, 4-(4-α-Cumylphenoxy)phthalonitrile, 2-[4-(3,4-Dicyanophenoxy)phenyl]-2-phenylpropane

A151599-1G A151599-250MG A151599-25G A151599-5G
ALA151599-1GEA 140.51 USD
ALA151599-1G ALA151599250MG ALA151599-25G ALA151599-5G
4-(4-ɑ-Cumylphenoxy)phthalonitrile ≥98%
4-(4-alpha-Cumylphenoxy)phthalonitrile
Formula: C₂₃H₁₈N₂O
MW: 338.41 g/mol
Melting Pt: 91 °C
MDL Number: MFCD00059048
CAS Number: 83482-57-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c907b881abc59e47f6d8e390e37937e25c13bacc&vid=c907b881abc59e47f6d8e390e37937e25c13bacc&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR