5-Amino-2-naphthalenesulfonic acid ≥97% (by titration)

Supplier: ALADDIN SCIENTIFIC
Warning

A151595-100G A151595-25G A151595-5G
ALA151595-100GEA 28.41 USD
ALA151595-100G ALA151595-25G ALA151595-5G
5-Amino-2-naphthalenesulfonic acid ≥97% (by titration)
5-Amino-2-naphthalenesulphonic acid

The crystal structure of 5-amino-2-naphthalenesulfonic acid (1,6-Cleve′s acid) depicted the presence of a sulfonate-aminium group zwitterion.5-Amino-2-naphthalenesulfonic acid (1,6-Cleve′s acid) was used in the synthesis of nitrate reductase and detection of nitrate reduction by anaerobic bacteria.

Formula: C₁₀H₉NO₃S
Melting Pt: >300°C
Storage Temperature: Ambient
MDL Number: MFCD00004030
CAS Number: 119-79-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c7cee872504a158dc89d5d19e2f541e821135551&vid=c7cee872504a158dc89d5d19e2f541e821135551&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR