5-Chloro-3-methylanthranilic acid ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A151591-100G A151591-25G A151591-5G
ALA151591-100GEA 195.61 USD
ALA151591-100G ALA151591-25G ALA151591-5G
5-Chloro-3-methylanthranilic acid ≥98%
5-Chloro-3-methylanthranilic acid
Formula: C₈H₈ClNO₂
Melting Pt: 238 °C
Storage Temperature: Ambient
MDL Number: MFCD02358895
CAS Number: 20776-67-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c86b8ceef365f37944e3311396aab550c83b7562&vid=c86b8ceef365f37944e3311396aab550c83b7562&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR