6-Methylanthranil acid ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A151590-1G A151590-25G A151590-5G
ALA151590-1GEA 17.96 USD
ALA151590-1G ALA151590-25G ALA151590-5G
6-Methylanthranil acid ≥98%
6-Methylanthranil acid

2-Amino-6-methylbenzoic acid reacts with cyanogen bromide to prepare 2-amino-5-methyl-benzo[d][1,3]oxazin-4-one.

Formula: C₈H₉NO₂
Melting Pt: 124-130°C
MDL Number: MFCD00007809
CAS Number: 4389-50-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7e58fe6376576a591676c078065297c85f6fe5d&vid=a7e58fe6376576a591676c078065297c85f6fe5d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR