4-Acetylphenyl trifluoromethanesulfonate ≥95% (by GC)

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: Trifluoromethanesulfonic acid 4-Acetylphenyl Ester, 4-Acetylphenyl Trifluoromethanesulphonate, 4-Acetylphenyl Triflate, Trifluoromethanesulphonic acid 4-Acetylphenyl Ester

A151589-1G A151589-5G
ALA151589-1GEA 17.96 USD
ALA151589-1G ALA151589-5G
4-Acetylphenyl trifluoromethanesulfonate ≥95% (by GC)
4-Acetylphenyl trifluoromethanesulfonate

4-Acetylphenyl trifluoromethanesulfonate (4-acetylphenyl triflate) is an aryl triflate. It undergoes palladium-catalyzed cross-coupling reaction with optically active (2-cyclohexenyl)difluorophenylsilane to form corresponding cross-coupled product.

Formula: C₉H₇F₃O₄S
MW: 268.21 g/mol
Boiling Pt: 75-76°C
Flash Pt: >113 °C
Storage Temperature: Ambient
MDL Number: MFCD00191706
CAS Number: 109613-00-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07b9f549a489505fe288bf50806dc7500629a53a&vid=07b9f549a489505fe288bf50806dc7500629a53a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR