4-Acetyl-6-tert-butyl-1,1-dimethylindan ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: 4-Acetyl-6-(tert-butyl)-1,1,-dimethylindane

A151586-25G A151586-5G
ALA151586-25GEA 134.81 USD
ALA151586-25G ALA151586-5G
4-Acetyl-6-tert-butyl-1,1-dimethylindan ≥98%
4-Acetyl-6-tert-butyl-1,1-dimethylindan
Formula: C₁₇H₂₄O
MW: 244.38 g/mol
Boiling Pt: 117°C
Melting Pt: 76-80°C
MDL Number: MFCD00046324
CAS Number: 13171-00-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd3e7052c4e50042393740422a5038de0e96cd57&vid=dd3e7052c4e50042393740422a5038de0e96cd57&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR