2-Amino-5-bromo-3-pyridinol ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2-Amino-5-bromo-3-hydroxypyridine

A151583-1G A151583-250MG A151583-5G
ALA151583-1GEA 34.11 USD
ALA151583-1G ALA151583250MG ALA151583-5G
2-Amino-5-bromo-3-pyridinol ≥98% (by HPLC)
2-Amino-5-bromo-3-pyridinol
Formula: C₅H₅BrN₂O
MW: 189.01 g/mol
Melting Pt: 204-207°C
MDL Number: MFCD09744143
CAS Number: 39903-01-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8375581aa4a974d21431e08f494364e6cf33c400&vid=8375581aa4a974d21431e08f494364e6cf33c400&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR