2-Amino-5-chloro-3-nitropyridine ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 5-Chloro-3-nitro-2-pyridylamine, 5-Chloro-3-nitropyridin-2-amine

A151579-100G A151579-25G A151579-5G
ALA151579-100GEA 555.66 USD
ALA151579-100G ALA151579-25G ALA151579-5G
2-Amino-5-chloro-3-nitropyridine ≥98%
2-Amino-5-chloro-3-nitropyridine
Formula: C₅H₄ClN₃O₂
MW: 173.56
Melting Pt: 190-195°C
MDL Number: MFCD00092011
CAS Number: 5409-39-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07c1b243b1325fb443e211169fc8b8a7aca06630&vid=07c1b243b1325fb443e211169fc8b8a7aca06630&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR