3-Amino-2-chloropyridine ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2-Chloro-3-pyridylamine, 2-Chloropyridin-3-amine

A151576-100G A151576-250G A151576-25G A151576-50G A151576-5G
ALA151576-100GEA 99.66 USD
ALA151576-100G ALA151576-250G ALA151576-25G ALA151576-50G ALA151576-5G
3-Amino-2-chloropyridine ≥98%
3-Amino-2-chloropyridine
Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Boiling Pt: 263
Melting Pt: 78 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006238
CAS Number: 6298-19-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a22ef2da8474768d85bb391c5a252a2d24fcc2db&vid=a22ef2da8474768d85bb391c5a252a2d24fcc2db&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR