Acetophenone-O-benzoyloxime 98

Supplier: ALADDIN SCIENTIFIC

Synonyms: Acetophenone O-Benzoyloxime

A151575-1G A151575-200MG A151575-5G
ALA151575-1GEA 100.61 USD
ALA151575-1G ALA151575200MG ALA151575-5G
Acetophenone-O-benzoyloxime 98
Acetophenone-O-benzoyloxime
Formula: C₁₅H₁₃NO₂
MW: 239.27 g/mol
Melting Pt: 100 °C
CAS Number: 26060-56-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0da9a88a991e7a6d503306fc5134c5482ad1a9d9&vid=0da9a88a991e7a6d503306fc5134c5482ad1a9d9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR