4-Anilinopyridine ≥99% (by titration)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: N-(4-Pyridyl)aniline, N-Phenylpyridin-4-amine

A151570-5G A151570-1G
ALA151570-5GEA 90.16 USD
ALA151570-5G ALA151570-1G
4-Anilinopyridine ≥99% (by titration)
4-Anilinopyridine
Formula: C₁₁H₁₀N₂
MW: 170.22 g/mol
Melting Pt: 177 °C
MDL Number: MFCD00457758
CAS Number: 22961-45-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=89b888b13e35fc445646d5d383477fac1b935a8c&vid=89b888b13e35fc445646d5d383477fac1b935a8c&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR