2-Amino-3-benzyloxypyridine ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: 3-Benzyloxypyridin-2-amine, 3-Benzyloxy-2-pyridylamine

A151569-25G A151569-5G
ALA151569-25GEA 99.66 USD
ALA151569-25G ALA151569-5G
2-Amino-3-benzyloxypyridine ≥98% (by GC)
2-Amino-3-benzyloxypyridine

2-Amino-3-benzyloxypyridine on condensation with diethyl ethoxymethylene malonate affords 9-benzyloxy-3-ethoxycarbonylpyrido[1,2-a]pyrimidin-4-one.2-Amino-3-benzyloxypyridine was used in the synthesis of 1-acetyl-2-[2-(3-benzyloxypyridinyl)]iminoimidazolidine.

Formula: C₁₂H₁₂N₂O
MW: 200.24 g/mol
Melting Pt: 97 °C
MDL Number: MFCD00006316
CAS Number: 24016-03-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=14cb73c124b5308c370b9589977572d40357b0d9&vid=14cb73c124b5308c370b9589977572d40357b0d9&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR