1-Aza-18-crown-6 (1,4,7,10,13-pentaoxa-16-azacyclooctadecane) ≥98% (by titration)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 1,4,7,10,13-Pentaoxa-16-azacyclooctadecane, 1-Aza-18-crown 6-Ether, 1-Aza-18-crown-6

A151566-1G A151566-250MG
ALA151566-1GEA 145.26 USD
ALA151566-1G ALA151566250MG
1-Aza-18-crown-6 (1,4,7,10,13-pentaoxa-16-azacyclooctadecane) ≥98% (by titration)
1-Aza-18-crown-6 (1,4,7,10,13-Pentaoxa-16-azacyclooctadecane)
Formula: C₁₂H₂₅NO₅
MW: 263.33 g/mol
Melting Pt: 51 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00075466
CAS Number: 33941-15-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f5ba89fbdc439fc710950a3a541abb78ef3d47f8&vid=f5ba89fbdc439fc710950a3a541abb78ef3d47f8&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR