2-Amino-5-chlorobenzonitrile ≥98% (by GC)
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Danger
|
Synonyms:
5-Chloroanthranilonitrile
A151558-1G
A151558-25G
A151558-5G
A151558-100G
ALA151558-1GEA
10.36
USD
ALA151558-1G
ALA151558-25G
ALA151558-5G
ALA151558-100G
2-Amino-5-chlorobenzonitrile ≥98% (by GC)
2-Amino-5-chlorobenzonitrile
The vibrational spectra of 2-amino-5-chlorobenzonitrile was studied using density functinal theory.2-Amino-5-chlorobenzonitrile was used to build up the 2,2-dioxide-1H-2,1,3-benzothiadiazine ring system.
Formula:
C₇H₅ClN₂ MW: 152.58 g/mol Boiling Pt: 159 °C Melting Pt: 96 °C |
MDL Number:
MFCD00017106 CAS Number: 5922-60-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...